Anti-AUH Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: AUH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77824-361
- 77824-360
- Antigen Symbol:
- AUH
- AUH
- Antigen Name:
- Methylglutaconyl-CoA hydratase, mitochondrial protein (68-339AA)
- Methylglutaconyl-CoA hydratase, mitochondrial protein (68-339AA)
- Antigen Synonyms:
- AUHMethylglutaconyl-CoA hydratase|mitochondrial|EC 4.2.1.18|AU-specific RNA-binding enoyl-CoA hydratase|AU-binding protein/enoyl-CoA hydratase|Itaconyl-CoA hydratase|EC 4.2.1.56
- AUHMethylglutaconyl-CoA hydratase|mitochondrial|EC 4.2.1.18|AU-specific RNA-binding enoyl-CoA hydratase|AU-binding protein/enoyl-CoA hydratase|Itaconyl-CoA hydratase|EC 4.2.1.56
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13825
- Q13825
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together