Anti-ABCA3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ABCA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-959
- 77834-960
- Antigen Symbol:
- ABCA3
- ABCA3
- Antigen Name:
- ATP-binding cassette sub-family A member 3 protein (43-203AA)
- ATP-binding cassette sub-family A member 3 protein (43-203AA)
- Antigen Synonyms:
- ABC 3|ABC C|ABC C transporter|ABC transporter 3|ABC-C transporter|ABC3|ABCA 3|Abca3|ABCA3 protein|ABCA3_HUMAN|ABCC|ATP binding cassette 3|ATP binding cassette sub family A (ABC1) member 3|ATP binding cassette sub family A member 3|ATP binding cassette transporter 3|ATP-binding cassette 3|ATP-binding cassette sub-family A member 3|ATP-binding cassette transporter 3|CED7. C. elegans|homolog of|EST111653|LBM 180|LBM180|MGC72201|P180 Lamellar Body Protein|SMDP3
- ABC 3|ABC C|ABC C transporter|ABC transporter 3|ABC-C transporter|ABC3|ABCA 3|Abca3|ABCA3 protein|ABCA3_HUMAN|ABCC|ATP binding cassette 3|ATP binding cassette sub family A (ABC1) member 3|ATP binding cassette sub family A member 3|ATP binding cassette transporter 3|ATP-binding cassette 3|ATP-binding cassette sub-family A member 3|ATP-binding cassette transporter 3|CED7. C. elegans|homolog of|EST111653|LBM 180|LBM180|MGC72201|P180 Lamellar Body Protein|SMDP3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99758
- Q99758
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together