Anti-RABEP2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: RABEP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-675
- 77840-676
- Antigen Symbol:
- RABEP2
- RABEP2
- Antigen Name:
- Rab GTPase-binding effector protein 2 protein (301-533AA)
- Rab GTPase-binding effector protein 2 protein (301-533AA)
- Antigen Synonyms:
- FLJ23282| FRA| Rab GTPase binding effector protein 2| Rab GTPase-binding effector protein 2| Rabaptin 5 beta| Rabaptin RAB GTPase binding effector protein 2| Rabaptin-5beta| RABE2_HUMAN| RABEP 2| | RABPT5B
- FLJ23282| FRA| Rab GTPase binding effector protein 2| Rab GTPase-binding effector protein 2| Rabaptin 5 beta| Rabaptin RAB GTPase binding effector protein 2| Rabaptin-5beta| RABE2_HUMAN| RABEP 2| | RABPT5B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H5N1
- Q9H5N1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together