Anti-CEP68 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CEP68
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-570
- 77831-569
- Antigen Symbol:
- CEP68
- CEP68
- Antigen Name:
- Centrosomal protein of 68 kDa protein (280-480AA)
- Centrosomal protein of 68 kDa protein (280-480AA)
- Antigen Synonyms:
- Centrosomal protein 68kDa|Centrosomal protein of 68 kDa|Cep68|CEP68_HUMAN|FLJ25920|FLJ36750|KIAA0582
- Centrosomal protein 68kDa|Centrosomal protein of 68 kDa|Cep68|CEP68_HUMAN|FLJ25920|FLJ36750|KIAA0582
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q76N32
- Q76N32
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together