Anti-SOCS7 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SOCS7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77800-622
- 77800-621
- Antigen Symbol:
- SOCS7
- SOCS7
- Antigen Name:
- Suppressor of cytokine signaling 7 protein (6-165AA)
- Suppressor of cytokine signaling 7 protein (6-165AA)
- Antigen Synonyms:
- Ash and phospholipase C gamma-binding protein|NAP 4|NAP 4 Fragment|NAP-4|NAP4|Nck|Nck Ash and phospholipase C binding protein|Nck Ash and phospholipase C gamma binding protein|Nck associated protein 4|Nck-associated protein 4|NCKAP4|SH2 domain containing SOCS box protein|SOCS 7|SOCS-7|SOCS4|SOCS6|Socs7|SOCS7_HUMAN|Suppressor of cytokine signaling 7
- Ash and phospholipase C gamma-binding protein|NAP 4|NAP 4 Fragment|NAP-4|NAP4|Nck|Nck Ash and phospholipase C binding protein|Nck Ash and phospholipase C gamma binding protein|Nck associated protein 4|Nck-associated protein 4|NCKAP4|SH2 domain containing SOCS box protein|SOCS 7|SOCS-7|SOCS4|SOCS6|Socs7|SOCS7_HUMAN|Suppressor of cytokine signaling 7
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14512
- O14512
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together