Anti-TAF8 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: TAF8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77832-620
- 77832-619
- Antigen Symbol:
- TAF8
- TAF8
- Antigen Name:
- Transcription initiation factor TFIID subunit 8 protein (122-311AA)
- Transcription initiation factor TFIID subunit 8 protein (122-311AA)
- Antigen Synonyms:
- FLJ32821|hTAFII43|Protein taube nuss|TAF(II)43|taf8|TAF8 RNA polymerase II, TATA box binding protein (TBP) associated factor, 43kDa|TAF8_HUMAN|TAFII-43|TAFII43|TATA box binding protein (TBP) associated factor, RNA polymerase II A 45/50kDa|Taube nuss homolog (mouse)|TBN|TBP associated factor 8|TBP associated factor TAFII43|TBP associated factor, RNA polymerase II, 43 kD|TBP-associated factor 43 kDa|TBP-associated factor 8|Transcription initiation factor TFIID 43 kDa subunit|Transcription initiation factor TFIID subunit 8
- FLJ32821|hTAFII43|Protein taube nuss|TAF(II)43|taf8|TAF8 RNA polymerase II, TATA box binding protein (TBP) associated factor, 43kDa|TAF8_HUMAN|TAFII-43|TAFII43|TATA box binding protein (TBP) associated factor, RNA polymerase II A 45/50kDa|Taube nuss homolog (mouse)|TBN|TBP associated factor 8|TBP associated factor TAFII43|TBP associated factor, RNA polymerase II, 43 kD|TBP-associated factor 43 kDa|TBP-associated factor 8|Transcription initiation factor TFIID 43 kDa subunit|Transcription initiation factor TFIID subunit 8
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z7C8
- Q7Z7C8
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together