Anti-TIPRL Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TIPRL
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77802-623
- 77802-624
- Antigen Symbol:
- TIPRL
- TIPRL
- Antigen Name:
- TIP41-like protein (1-178AA)
- TIP41-like protein (1-178AA)
- Antigen Synonyms:
- 1810011K17Rik|dJ69E11.3|MGC3794|Putative MAPK activating protein PM10|Putative MAPK-activating protein PM10|RP1-69E11.1|Tip|TIP41|TIP41-like protein|tiprl|TIPRL_HUMAN|TOR signaling pathway regulator|TOR Signaling Pathway Regulator like|Type 2A-interacting protein
- 1810011K17Rik|dJ69E11.3|MGC3794|Putative MAPK activating protein PM10|Putative MAPK-activating protein PM10|RP1-69E11.1|Tip|TIP41|TIP41-like protein|tiprl|TIPRL_HUMAN|TOR signaling pathway regulator|TOR Signaling Pathway Regulator like|Type 2A-interacting protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75663
- O75663
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together