Anti-GCSAM Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GCSAM
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-369
- 77833-368
- Antigen Symbol:
- GCSAM
- GCSAM
- Antigen Name:
- Germinal center-associated signaling and motility protein (1-178AA)
- Germinal center-associated signaling and motility protein (1-178AA)
- Antigen Synonyms:
- GCAT2|Gcet|GCET2 germinal center expressed transcript 2|GCET2 germinal centre expressed transcript 2|Gcsam|GCSAM_HUMAN|Germinal center associated lymphoma |Germinal center associated lymphoma protein|Germinal center B cell associated protein 2|Germinal center B-cell-expressed transcript 2 protein|Germinal center expressed transcript 2|Germinal center-associated lymphoma protein|Germinal center-associated signaling and motility protein|Germinal centre associated lymphoma |Germinal centre associated lymphoma protein|Germinal centre B cell associated protein 2|Germinal centre expressed transcript 2|hGAL|M17|M17 L
- GCAT2|Gcet|GCET2 germinal center expressed transcript 2|GCET2 germinal centre expressed transcript 2|Gcsam|GCSAM_HUMAN|Germinal center associated lymphoma |Germinal center associated lymphoma protein|Germinal center B cell associated protein 2|Germinal center B-cell-expressed transcript 2 protein|Germinal center expressed transcript 2|Germinal center-associated lymphoma protein|Germinal center-associated signaling and motility protein|Germinal centre associated lymphoma |Germinal centre associated lymphoma protein|Germinal centre B cell associated protein 2|Germinal centre expressed transcript 2|hGAL|M17|M17 L
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N6F7
- Q8N6F7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together