Anti-CHP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CHP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-113
- 77835-112
- Antigen Symbol:
- CHP1
- CHP1
- Antigen Name:
- Calcineurin B homologous protein 1 protein (2-195AA)
- Calcineurin B homologous protein 1 protein (2-195AA)
- Antigen Synonyms:
- Calcineurin B homolog|Calcineurin B homologous protein|Calcineurin homologous protein|Calcineurin like EF hand protein 1|Calcium binding protein CHP|Calcium binding protein p22|Calcium-binding protein CHP|Calcium-binding protein p22|chp|CHP1_HUMAN|Sid 470|Sid470|SLC9A1 binding protein|SLC9A1BP|Wrch2
- Calcineurin B homolog|Calcineurin B homologous protein|Calcineurin homologous protein|Calcineurin like EF hand protein 1|Calcium binding protein CHP|Calcium binding protein p22|Calcium-binding protein CHP|Calcium-binding protein p22|chp|CHP1_HUMAN|Sid 470|Sid470|SLC9A1 binding protein|SLC9A1BP|Wrch2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99653
- Q99653
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together