Anti-DCTN2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DCTN2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77822-425
- 77822-426
- Antigen Symbol:
- DCTN2
- DCTN2
- Antigen Name:
- Dynactin subunit 2 protein (1-252AA)
- Dynactin subunit 2 protein (1-252AA)
- Antigen Synonyms:
- 50 kD dynein associated polypeptide |50 kDa dynein associated polypeptide |50 kDa dynein-associated polypeptide|DCTN-50||_HUMAN|DCTN50 |dynactin 2 (p50) |dynactin 2|dynactin complex 50 kD subunit |Dynactin complex 50 kDa subunit|Dynactin subunit 2|DYNAMITIN|p50 dynamitin|RBP50
- 50 kD dynein associated polypeptide |50 kDa dynein associated polypeptide |50 kDa dynein-associated polypeptide|DCTN-50||_HUMAN|DCTN50 |dynactin 2 (p50) |dynactin 2|dynactin complex 50 kD subunit |Dynactin complex 50 kDa subunit|Dynactin subunit 2|DYNAMITIN|p50 dynamitin|RBP50
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13561
- Q13561
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together