Anti-RBFOX3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RBFOX3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77818-136
- 77818-135
- Antigen Symbol:
- RBFOX3
- RBFOX3
- Antigen Name:
- RNA binding protein fox-1 homolog 3 protein (1-101AA)
- RNA binding protein fox-1 homolog 3 protein (1-101AA)
- Antigen Synonyms:
- FLJ56884| FLJ58356| Fox-1 homolog C| fox1 homolog C| Fox3| FOX3NeuN| hexaribonucleotide binding protein 3| HRNBP3| NEUN| neuronal nuclei| Rbfox3| RFOX3_HUMAN| RNA binding protein fox-1 homolog 3| RNA binding protein, fox 1 homolog (C. elegans) 3
- FLJ56884| FLJ58356| Fox-1 homolog C| fox1 homolog C| Fox3| FOX3NeuN| hexaribonucleotide binding protein 3| HRNBP3| NEUN| neuronal nuclei| Rbfox3| RFOX3_HUMAN| RNA binding protein fox-1 homolog 3| RNA binding protein, fox 1 homolog (C. elegans) 3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A6NFN3
- A6NFN3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together