Anti-TPP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TPP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-387
- 77803-387
- Antigen Symbol:
- TPP1
- TPP1
- Antigen Name:
- Tripeptidyl-peptidase 1 protein (371-480AA)
- Tripeptidyl-peptidase 1 protein (371-480AA)
- Antigen Synonyms:
- Cell growth inhibiting gene 1 protein|Cell growth-inhibiting gene 1 protein|Ceroid lipofuscinosis neuronal 2|Ceroid lipofuscinosis neuronal 2 late infantile (Jansky Bielschowsky disease)|Ceroid lipofuscinosis neuronal 2 late infantile|CLN 2|CLN2|GIG 1|GIG1|Growth inhibiting protein 1|LPIC|Lysosomal pepstatin insensitive protease|Lysosomal pepstatin-insensitive protease|MGC21297|TPP 1|TPP I|TPP-1|TPP-I|Tpp1|TPP1_HUMAN|TPPI|Tripeptidyl aminopeptidase|Tripeptidyl peptidase I|Tripeptidyl-peptidase 1|Tripeptidyl-peptidase I
- Cell growth inhibiting gene 1 protein|Cell growth-inhibiting gene 1 protein|Ceroid lipofuscinosis neuronal 2|Ceroid lipofuscinosis neuronal 2 late infantile (Jansky Bielschowsky disease)|Ceroid lipofuscinosis neuronal 2 late infantile|CLN 2|CLN2|GIG 1|GIG1|Growth inhibiting protein 1|LPIC|Lysosomal pepstatin insensitive protease|Lysosomal pepstatin-insensitive protease|MGC21297|TPP 1|TPP I|TPP-1|TPP-I|Tpp1|TPP1_HUMAN|TPPI|Tripeptidyl aminopeptidase|Tripeptidyl peptidase I|Tripeptidyl-peptidase 1|Tripeptidyl-peptidase I
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14773
- O14773
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together