Anti-DCLK2 Rabbit Polyclonal Antibody
Catalog # 77813-671
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of DCLK2.
- Antigen Symbol:DCLK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q8N568
- Antigen Synonyms:CaMK-like CREB regulatory kinase 2|CL2|CLICK-II|CLICK2|DCAMK L2|DCAMKL 2|DCAMKL2|DCDC 3|DCDC 3B|DCDC3|DCDC3B|DCK 2|DCK2|DCLK 2|Dclk2|DCLK2_HUMAN|DKFZp761I032|Doublecortin and CaM kinase like 2|Doublecortin domain-containing protein 3B|Doublecortin like and CAM kinase like 2|Doublecortin like kinase 2|Doublecortin-like and CAM kinase-like 2|Doublecortin-like kinase 2|MGC45428|Serine/threonine protein kinase DCLK2|Serine/threonine-protein kinase DCLK2
- Storage Buffer:PBS, 150mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: DCLK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse