Anti-PDZD3 Rabbit Recombinant Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PDZD3
Clonality: Recombinant
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-687
- 77832-688
- Antigen Symbol:
- PDZD3
- PDZD3
- Antigen Name:
- Na(+)/H(+) exchange regulatory cofactor NHE-RF4 protein (42-291AA)
- Na(+)/H(+) exchange regulatory cofactor NHE-RF4 protein (42-291AA)
- Antigen Synonyms:
- IKEPP|Intestinal and kidney enriched PDZ protein|Intestinal and kidney-enriched PDZ protein|Na(+)/H(+) exchange regulatory cofactor NHE-RF4|Na/Pi cotransporter C terminal associated protein 2|Na/Pi cotransporter C-terminal-associated protein 2|NaPi-Cap2|Natrium-phosphate cotransporter IIa C-terminal-associated protein 2|NHERF-4|NHERF4|NHRF4_HUMAN|PDZ domain containing 2|PDZ domain containing 3|PDZ domain containing protein 2|PDZ domain-containing protein 2|PDZ domain-containing protein 3|PDZD 3|Pdzd3|PDZK2|Protein DLNB27|Sodium-hydrogen exchanger regulatory factor 4
- IKEPP|Intestinal and kidney enriched PDZ protein|Intestinal and kidney-enriched PDZ protein|Na(+)/H(+) exchange regulatory cofactor NHE-RF4|Na/Pi cotransporter C terminal associated protein 2|Na/Pi cotransporter C-terminal-associated protein 2|NaPi-Cap2|Natrium-phosphate cotransporter IIa C-terminal-associated protein 2|NHERF-4|NHERF4|NHRF4_HUMAN|PDZ domain containing 2|PDZ domain containing 3|PDZ domain containing protein 2|PDZ domain-containing protein 2|PDZ domain-containing protein 3|PDZD 3|Pdzd3|PDZK2|Protein DLNB27|Sodium-hydrogen exchanger regulatory factor 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86UT5
- Q86UT5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together