Anti-CENPX Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CENPX
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-402
- 77801-401
- Antigen Symbol:
- CENPX
- CENPX
- Antigen Name:
- Centromere protein X protein (1-63AA)
- Centromere protein X protein (1-63AA)
- Antigen Synonyms:
- CENP X|CENP-X|CENPX|CENPX_HUMAN|Centromere protein X|FAAP10|FANCM interacting histone fold protein 2|FANCM-interacting histone fold protein 2|Fanconi anemia associated polypeptide of 10 kDa|Fanconi anemia-associated polypeptide of 10 kDa|MGC14480|Retinoic acid inducible gene D9 protein homolog|Retinoic acid-inducible gene D9 protein homolog|Stimulated by retinoic acid gene 13 protein homolog|STRA13
- CENP X|CENP-X|CENPX|CENPX_HUMAN|Centromere protein X|FAAP10|FANCM interacting histone fold protein 2|FANCM-interacting histone fold protein 2|Fanconi anemia associated polypeptide of 10 kDa|Fanconi anemia-associated polypeptide of 10 kDa|MGC14480|Retinoic acid inducible gene D9 protein homolog|Retinoic acid-inducible gene D9 protein homolog|Stimulated by retinoic acid gene 13 protein homolog|STRA13
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A8MT69
- A8MT69
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together