Anti-SPC25 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SPC25
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-793
- 77840-794
- Antigen Symbol:
- SPC25
- SPC25
- Antigen Name:
- Kinetochore protein Spc25 protein (1-224AA)
- Kinetochore protein Spc25 protein (1-224AA)
- Antigen Synonyms:
- AD024|hSpc25|Kinetochore protein Spc25|SPBC25|SPC25|SPC25_HUMAN|spindle pole body component 25 homolog
- AD024|hSpc25|Kinetochore protein Spc25|SPBC25|SPC25|SPC25_HUMAN|spindle pole body component 25 homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HBM1
- Q9HBM1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together