Anti-UPF2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: UPF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-265
- 77837-264
- Antigen Symbol:
- UPF2
- UPF2
- Antigen Name:
- Regulator of nonsense transcripts 2 protein (1153-1272AA)
- Regulator of nonsense transcripts 2 protein (1153-1272AA)
- Antigen Synonyms:
- DKFZp434D222|hUpf2|KIAA1408|MGC138834|MGC138835|Nonsense mRNA reducing factor 2|Regulator of nonsense transcripts 2|RENT2|RENT2_HUMAN|Smg 3|Smg 3 homolog nonsense mediated mRNA decay factor|Up-frameshift suppressor 2 homolog|UPF2|UPF2 regulator of nonsense transcripts homolog|UROC1|Urocanate hydratase|Yeast Upf2p homolog
- DKFZp434D222|hUpf2|KIAA1408|MGC138834|MGC138835|Nonsense mRNA reducing factor 2|Regulator of nonsense transcripts 2|RENT2|RENT2_HUMAN|Smg 3|Smg 3 homolog nonsense mediated mRNA decay factor|Up-frameshift suppressor 2 homolog|UPF2|UPF2 regulator of nonsense transcripts homolog|UROC1|Urocanate hydratase|Yeast Upf2p homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HAU5
- Q9HAU5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together