Anti-ARMCX2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ARMCX2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77830-783
- 77830-784
- Antigen Symbol:
- ARMCX2
- ARMCX2
- Antigen Name:
- Armadillo repeat-containing X-linked protein 2 protein (161-460AA)
- Armadillo repeat-containing X-linked protein 2 protein (161-460AA)
- Antigen Synonyms:
- ALEX 2|ALEX2|ARM protein lost in epithelial cancers on chromosome X 2|Armadillo repeat containing X linked protein 2|Armadillo repeat containing, X linked 2|Armadillo repeat protein ALEX2|Armadillo repeat-containing X-linked protein 2|ARMCX 2|ARMCX2|ARMX2_HUMAN|GASP9|KIAA0512|MGC13343|MGC8742|Protein ALEX2
- ALEX 2|ALEX2|ARM protein lost in epithelial cancers on chromosome X 2|Armadillo repeat containing X linked protein 2|Armadillo repeat containing, X linked 2|Armadillo repeat protein ALEX2|Armadillo repeat-containing X-linked protein 2|ARMCX 2|ARMCX2|ARMX2_HUMAN|GASP9|KIAA0512|MGC13343|MGC8742|Protein ALEX2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7L311
- Q7L311
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together