Anti-TRIM49 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:30 to 1:150
Type:
Antigen: TRIM49
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77821-455
- 77821-456
- Antigen Symbol:
- TRIM49
- TRIM49
- Antigen Name:
- Fusion protein of TRIM49
- Fusion protein of TRIM49
- Antigen Synonyms:
- RING finger protein 18|RNF 18|RNF18|Testis specific RING Finger protein|Testis-specific ring-finger protein|TRI49_HUMAN|TRIM 49|TRIM49|Tripartite motif containing 49|Tripartite motif containing protein 49|Tripartite motif-containing protein 49
- RING finger protein 18|RNF 18|RNF18|Testis specific RING Finger protein|Testis-specific ring-finger protein|TRI49_HUMAN|TRIM 49|TRIM49|Tripartite motif containing 49|Tripartite motif containing protein 49|Tripartite motif-containing protein 49
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0CI25
- P0CI25
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together