Anti-TMEM68 Rabbit Recombinant Polyclonal Antibody
Catalog # 77803-117
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Transmembrane protein 68 protein (74-114AA)
- Antigen Symbol:TMEM68
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96MH6
- Antigen Synonyms:2010300G19Rik|AA408325|AW123402|RGD1309006|RP23-196C23.3|TMEM68|TMM68_HUMAN|Transmembrane protein 68
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: TMEM68
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse