Anti-MYH14 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYH14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-559
- 77832-558
- Antigen Symbol:
- MYH14
- MYH14
- Antigen Name:
- Myosin-14 protein (1058-1195AA)
- Myosin-14 protein (1058-1195AA)
- Antigen Synonyms:
- 2400004E04Rik|DFNA4|DKFZp667A1311|FLJ13881|FLJ43092|FP17425|II C|KIAA2034|MHC16|Myh 14|MYH14|MYH14_HUMAN|Myosin 14|Myosin|Myosin heavy chain 14|Myosin heavy chain|Myosin heavy chain non muscle IIc|Myosin heavy polypeptide 14|Myosin-14|NMHC II C|NMHC II-C|Non muscle myosin heavy chain IIc|non-muscle IIc|Non-muscle myosin heavy chain IIc|Nonmuscle myosin heavy chain II C|OTTMUSP00000019210
- 2400004E04Rik|DFNA4|DKFZp667A1311|FLJ13881|FLJ43092|FP17425|II C|KIAA2034|MHC16|Myh 14|MYH14|MYH14_HUMAN|Myosin 14|Myosin|Myosin heavy chain 14|Myosin heavy chain|Myosin heavy chain non muscle IIc|Myosin heavy polypeptide 14|Myosin-14|NMHC II C|NMHC II-C|Non muscle myosin heavy chain IIc|non-muscle IIc|Non-muscle myosin heavy chain IIc|Nonmuscle myosin heavy chain II C|OTTMUSP00000019210
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z406
- Q7Z406
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together