Anti-RAP2B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: RAP2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77796-656
- 77796-655
- Antigen Symbol:
- RAP2B
- RAP2B
- Antigen Name:
- Ras-related protein Rap-2b protein (1-183AA)
- Ras-related protein Rap-2b protein (1-183AA)
- Antigen Synonyms:
- MGC20484|RAP 2B|RAP2A|Rap2b|RAP2B member of RAS oncogene family|RAP2B_HUMAN|Ras family small GTP binding protein RAP2B|Ras related protein RAP 2B|Ras related protein RAP2B|Ras-related protein Rap-2b|Small GTP binding protein
- MGC20484|RAP 2B|RAP2A|Rap2b|RAP2B member of RAS oncogene family|RAP2B_HUMAN|Ras family small GTP binding protein RAP2B|Ras related protein RAP 2B|Ras related protein RAP2B|Ras-related protein Rap-2b|Small GTP binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P61225
- P61225
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together