Anti-mutT Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: mutT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli
Specifications
- Catalog No:
- 77817-084
- 77817-085
- Antigen Symbol:
- mutT
- mutT
- Antigen Name:
- Recombinant Escherichia coli 8-oxo-dGTP diphosphatase protein (1-129AA)
- Recombinant Escherichia coli 8-oxo-dGTP diphosphatase protein (1-129AA)
- Antigen Synonyms:
- mutT|b0099|JW0097|8-oxo-dGTP diphosphatase|8-oxo-dGTPase|EC 3.6.1.55|7,8-dihydro-8-oxoguanine-triphosphatase|Mutator protein MutT|dGTP pyrophosphohydrolase
- mutT|b0099|JW0097|8-oxo-dGTP diphosphatase|8-oxo-dGTPase|EC 3.6.1.55|7,8-dihydro-8-oxoguanine-triphosphatase|Mutator protein MutT|dGTP pyrophosphohydrolase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- E. coli
- E. coli
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P08337
- P08337
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together