Anti-VTN Rabbit Polyclonal Antibody
Catalog # 77804-458
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:VTNC_HUMAN
- Antigen Symbol:VTN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P04004
- Antigen Synonyms:Complement S Protein|Epibolin|S Protein|S-protein|Serum Spreading Factor|Serum-spreading factor|Somatomedin B|Somatomedin-B|V75|Vitronectin|Vitronectin V10 subunit|Vitronectin V65 subunit|VN|VNT|VTN|VTNC_HUMAN
- Storage Buffer:PBS, 0.1% Sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: VTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat