Anti-EAF2 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: EAF2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77833-993
- 77833-992
- Antigen Symbol:
- EAF2
- EAF2
- Antigen Name:
- ELL-associated factor 2 protein (6-100AA)
- ELL-associated factor 2 protein (6-100AA)
- Antigen Synonyms:
- BM 040|BM040|EAF 2||_HUMAN|Ehrlich S II transcriptional activator factor |ELL associated factor 2|ELL-associated factor 2|Festa|Testosterone regulated apoptosis inducer and tumor suppressor|Testosterone regulated apoptosis inducer and tumor suppressor protein|Testosterone-regulated apoptosis inducer and tumor suppressor protein|TRAITS|U19|Uncharacterized bone marrow protein BM 040|Uncharacterized bone marrow protein BM040
- BM 040|BM040|EAF 2||_HUMAN|Ehrlich S II transcriptional activator factor |ELL associated factor 2|ELL-associated factor 2|Festa|Testosterone regulated apoptosis inducer and tumor suppressor|Testosterone regulated apoptosis inducer and tumor suppressor protein|Testosterone-regulated apoptosis inducer and tumor suppressor protein|TRAITS|U19|Uncharacterized bone marrow protein BM 040|Uncharacterized bone marrow protein BM040
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96CJ1
- Q96CJ1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together