Anti-ACO1 Rabbit Polyclonal Antibody
77797-815
New Product
- Antigen Name:Synthesized peptide derived from IRP-1 around the non-phosphorylation site of S138
- Antigen Symbol:ACO1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P21399
- Antigen Synonyms:ACO 1|ACO1|ACOC_HUMAN|Aconitase 1 soluble|Aconitase|Aconitase1|Aconitate hydratase|ACONS|Citrate hydro lyase|Citrate hydro-lyase|Cytoplasmic aconitate hydratase|Ferritin repressor protein|IRE BP 1|IRE-BP 1|IREB 1|IREB1|IREBP|IREBP1|Iron regulatory protein 1|Iron responsive element binding protein 1|Iron-responsive element-binding protein 1|IRP 1|IRP1|OTTHUMP00000045233
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% Sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended Dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:20000
Type:
Antigen: ACO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat