Anti-EXTL3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EXTL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-464
- 77806-463
- Antigen Symbol:
- EXTL3
- EXTL3
- Antigen Name:
- Exostosin-like 3 protein (2-245AA)
- Exostosin-like 3 protein (2-245AA)
- Antigen Synonyms:
- botv|DKFZp686C2342|Exostoses (multiple)-like 3|Exostoses-like 3|Exostosin-like 3|EXT-related protein 1|EXTL1L|EXTL3|EXTL3_HUMAN|EXTR1|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferas|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase|Hereditary multiple exostoses gene isolog|KIAA0519|Multiple exostosis-like protein 3|Putative tumor suppressor protein EXTL3|REG|Reg receptor|REGR|RPR
- botv|DKFZp686C2342|Exostoses (multiple)-like 3|Exostoses-like 3|Exostosin-like 3|EXT-related protein 1|EXTL1L|EXTL3|EXTL3_HUMAN|EXTR1|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferas|Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase|Hereditary multiple exostoses gene isolog|KIAA0519|Multiple exostosis-like protein 3|Putative tumor suppressor protein EXTL3|REG|Reg receptor|REGR|RPR
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43909
- O43909
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together