Anti-NUGGC Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: NUGGC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-108
- 77828-107
- Antigen Symbol:
- NUGGC
- NUGGC
- Antigen Name:
- Nuclear GTPase SLIP-GC protein (1-228AA)
- Nuclear GTPase SLIP-GC protein (1-228AA)
- Antigen Synonyms:
- NUGGC|C8orf80|HMFN0672|Nuclear GTPase SLIP-GC|EC 3.6.1.-|Speckled-like pattern in the germinal center
- NUGGC|C8orf80|HMFN0672|Nuclear GTPase SLIP-GC|EC 3.6.1.-|Speckled-like pattern in the germinal center
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q68CJ6
- Q68CJ6
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together