Anti-EEF1B2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EEF1B2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77816-076
- 77816-077
- Antigen Symbol:
- EEF1B2
- EEF1B2
- Antigen Name:
- Elongation factor 1-beta protein (1-225AA)
- Elongation factor 1-beta protein (1-225AA)
- Antigen Synonyms:
- EEF1B|EEF1B1|EEF1B2|EF-1-beta|EF1B|EF1B_HUMAN|Elongation factor 1|beta-2-A|Elongation factor 1-beta|eukaryotic translation elongation factor 1 beta 1|eukaryotic translation elongation factor 1 beta 2
- EEF1B|EEF1B1|EEF1B2|EF-1-beta|EF1B|EF1B_HUMAN|Elongation factor 1|beta-2-A|Elongation factor 1-beta|eukaryotic translation elongation factor 1 beta 1|eukaryotic translation elongation factor 1 beta 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P24534
- P24534
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together