Anti-RGS19 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: RGS19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77797-200
- 77797-199
- Antigen Symbol:
- RGS19
- RGS19
- Antigen Name:
- Regulator of G-protein signaling 19 protein (1-87AA)
- Regulator of G-protein signaling 19 protein (1-87AA)
- Antigen Synonyms:
- G alpha interacting protein|G protein signalling regulator 19|G protein|alpha-interacting protein|G-alpha-interacting protein|GAIP|GNAI3IP|Guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein|Regulator of G protein signalling 19|Regulator of G-protein signaling 19|RGS19|RGS19_HUMAN|RGSGAIP
- G alpha interacting protein|G protein signalling regulator 19|G protein|alpha-interacting protein|G-alpha-interacting protein|GAIP|GNAI3IP|Guanine nucleotide binding protein alpha inhibiting activity polypeptide 3 interacting protein|Regulator of G protein signalling 19|Regulator of G-protein signaling 19|RGS19|RGS19_HUMAN|RGSGAIP
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P49795
- P49795
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together