Anti-TTC19 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: TTC19
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77828-515
- 77828-514
- Antigen Symbol:
- TTC19
- TTC19
- Antigen Name:
- Tetratricopeptide repeat protein 19, mitochondrial protein (108-380AA)
- Tetratricopeptide repeat protein 19, mitochondrial protein (108-380AA)
- Antigen Synonyms:
- 2010204O13Rik|FLJ20343|MGC138312|MGC19520|mitochondrial|RIKEN cDNA 2810460C24|Tetratricopeptide repeat protein 19|TPR repeat protein 19|Ttc 19|TTC19|TTC19_HUMAN
- 2010204O13Rik|FLJ20343|MGC138312|MGC19520|mitochondrial|RIKEN cDNA 2810460C24|Tetratricopeptide repeat protein 19|TPR repeat protein 19|Ttc 19|TTC19|TTC19_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6DKK2
- Q6DKK2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together