Anti-DNM1 Rabbit Polyclonal Antibody
Catalog # 77808-987
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Dynamin-1
- Antigen Symbol:DNM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q05193
- Antigen Synonyms:B dynamin|D100|DNM 1|DNM|DNM1|DYN1_HUMAN|Dynamin|Dynamin-1|Dynamin1
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:1000
Type:
Antigen: DNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat