Anti-SNAPC1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SNAPC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77825-058
- 77825-059
- Antigen Symbol:
- SNAPC1
- SNAPC1
- Antigen Name:
- snRNA-activating protein complex subunit 1 protein (151-368AA)
- snRNA-activating protein complex subunit 1 protein (151-368AA)
- Antigen Synonyms:
- Proximal sequence element-binding transcription factor subunit gamma|PSE binding factor subunit gamma|PSE-binding factor subunit gamma|PTF subunit gamma|PTFgamma|Small nuclear RNA activating complex polypeptide 1|small nuclear RNA activating complex, polypeptide 1, 43kDa|Small nuclear RNA-activating complex polypeptide 1|Small nuclear RNA-activating protein complex, 43-KD subunit|SNAP43|SNAPc 43 kDa subunit|SNAPc subunit 1|Snapc1|SNPC1_HUMAN|snRNA activating protein complex 43 kDa subunit|snRNA activating protein complex subunit 1|snRNA-activating protein complex 43 kDa subunit|snRNA-activating protein complex subunit 1
- Proximal sequence element-binding transcription factor subunit gamma|PSE binding factor subunit gamma|PSE-binding factor subunit gamma|PTF subunit gamma|PTFgamma|Small nuclear RNA activating complex polypeptide 1|small nuclear RNA activating complex, polypeptide 1, 43kDa|Small nuclear RNA-activating complex polypeptide 1|Small nuclear RNA-activating protein complex, 43-KD subunit|SNAP43|SNAPc 43 kDa subunit|SNAPc subunit 1|Snapc1|SNPC1_HUMAN|snRNA activating protein complex 43 kDa subunit|snRNA activating protein complex subunit 1|snRNA-activating protein complex 43 kDa subunit|snRNA-activating protein complex subunit 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16533
- Q16533
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together