Anti-EGLN3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: EGLN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-703
- 77840-704
- Antigen Symbol:
- EGLN3
- EGLN3
- Antigen Name:
- Egl nine homolog 3 protein (1-120AA)
- Egl nine homolog 3 protein (1-120AA)
- Antigen Synonyms:
- Egl 9 family hypoxia inducible factor 3|Egl nine homolog 3 (C. elegans)|Egl nine homolog 3|Egl nine like protein 3 isoform|EGL9 homolog of C. elegans 3|EGLN3|EGLN3_HUMAN|Factor responsive smooth muscle protein|HIF Prolyl Hydroxylase 3|HIF-PH3|HIF-prolyl hydroxylase 3|HIFP4H3|HIFPH3|HPH-1|HPH-3|Hypoxia-inducible factor prolyl hydroxylase 3|P4H3|PHD3|Prolyl Hydroxylase Domain Containing Protein 3|Prolyl hydroxylase domain-containing protein 3|SM20
- Egl 9 family hypoxia inducible factor 3|Egl nine homolog 3 (C. elegans)|Egl nine homolog 3|Egl nine like protein 3 isoform|EGL9 homolog of C. elegans 3|EGLN3|EGLN3_HUMAN|Factor responsive smooth muscle protein|HIF Prolyl Hydroxylase 3|HIF-PH3|HIF-prolyl hydroxylase 3|HIFP4H3|HIFPH3|HPH-1|HPH-3|Hypoxia-inducible factor prolyl hydroxylase 3|P4H3|PHD3|Prolyl Hydroxylase Domain Containing Protein 3|Prolyl hydroxylase domain-containing protein 3|SM20
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H6Z9
- Q9H6Z9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together