Anti-aglB Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: aglB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Aspergillus niger
- Catalog No:
- 77817-974
- 77817-973
- Antigen Symbol:
- aglB
- aglB
- Antigen Name:
- Recombinant Aspergillus niger Probable alpha-galactosidase B protein (21-443AA)
- Recombinant Aspergillus niger Probable alpha-galactosidase B protein (21-443AA)
- Antigen Synonyms:
- An02g11150Probable alpha-galactosidase B|EC 3.2.1.22|Melibiase B
- An02g11150Probable alpha-galactosidase B|EC 3.2.1.22|Melibiase B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Aspergillus niger
- Aspergillus niger
- Host:
- Rabbit
- Rabbit
- Gene ID:
- A2QEJ9
- A2QEJ9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together