Anti-HDHD2 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: HDHD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77840-525
- 77840-526
- Antigen Symbol:
- HDHD2
- HDHD2
- Antigen Name:
- Haloacid dehalogenase-like hydrolase domain-containing protein 2 protein (97-249AA)
- Haloacid dehalogenase-like hydrolase domain-containing protein 2 protein (97-249AA)
- Antigen Synonyms:
- 0610039H12Rik|3110052N05Rik|DKFZP564D1378|Haloacid dehalogenase like hydrolase domain containing 2|Haloacid dehalogenase-like hydrolase domain-containing protein 2|HDHD2|HDHD2_HUMAN|Ier3ip1|Immediate early response 3 interacting protein 1|LRRG00122|OTTMUSP00000045339|OTTMUSP00000045340|OTTMUSP00000045341|OTTMUSP00000045343|OTTMUSP00000045344|RGD1308579
- 0610039H12Rik|3110052N05Rik|DKFZP564D1378|Haloacid dehalogenase like hydrolase domain containing 2|Haloacid dehalogenase-like hydrolase domain-containing protein 2|HDHD2|HDHD2_HUMAN|Ier3ip1|Immediate early response 3 interacting protein 1|LRRG00122|OTTMUSP00000045339|OTTMUSP00000045340|OTTMUSP00000045341|OTTMUSP00000045343|OTTMUSP00000045344|RGD1308579
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H0R4
- Q9H0R4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together