Order Entry
United States
ContactUsLinkComponent
Anti-EPN3 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Anti-EPN3 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-EPN3 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77837-107
  • 77837-106
  • Antigen Symbol:
  • EPN3
  • EPN3
  • Antigen Name:
  • Epsin-3 protein (25-208AA)
  • Epsin-3 protein (25-208AA)
  • Antigen Synonyms:
  • EPN 3|Epn3|EPN3_HUMAN|EPS 15 interacting protein 3|EPS-15-interacting protein 3|Epsin 3|Epsin-3|Epsin3|FLJ20778|MGC129899
  • EPN 3|Epn3|EPN3_HUMAN|EPS 15 interacting protein 3|EPS-15-interacting protein 3|Epsin 3|Epsin-3|Epsin3|FLJ20778|MGC129899
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9H201
  • Q9H201
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together