Anti-TADA3 Rabbit Recombinant Polyclonal Antibody
New Product
Type:
Antigen: TADA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77806-807
- 77806-806
- Antigen Symbol:
- TADA3
- TADA3
- Antigen Name:
- Transcriptional adapter 3 protein (1-238AA)
- Transcriptional adapter 3 protein (1-238AA)
- Antigen Synonyms:
- ADA3|TADA3L|Transcriptional adapter 3|ADA3 homolog|hADA3|STAF54|Transcriptional adapter 3-like|ADA3-like protein
- ADA3|TADA3L|Transcriptional adapter 3|ADA3 homolog|hADA3|STAF54|Transcriptional adapter 3-like|ADA3-like protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75528
- O75528
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together