Anti-Snrnp35 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: WB: 1:1000 to 1:5000
Type:
Antigen: Snrnp35
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- 77841-869
- 77841-870
- Antigen Symbol:
- Snrnp35
- Snrnp35
- Antigen Name:
- Recombinant Mouse U11/U12 small nuclear ribonucleoprotein 35 kDa protein (1-244AA)
- Recombinant Mouse U11/U12 small nuclear ribonucleoprotein 35 kDa protein (1-244AA)
- Antigen Synonyms:
- Snrnp35|U1snrnpbpU11/U12 small nuclear ribonucleoprotein 35 kDa protein|U11/U12 snRNP 35 kDa protein|U1 snRNP-binding protein homolog
- Snrnp35|U1snrnpbpU11/U12 small nuclear ribonucleoprotein 35 kDa protein|U11/U12 snRNP 35 kDa protein|U1 snRNP-binding protein homolog
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse, Human
- Mouse, Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9D384
- Q9D384
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together