Anti-KCNT1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KCNT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-930
- 77826-931
- Antigen Symbol:
- bA100C15.2;EIEE14;ENFL5;KCa4.1;KCNT1;KCNT1_HUMAN;Potassium channel subfamily T member 1;Potassium channel, sodium activated subfamily T, member 1;Potassium channel, subfamily T, member 1;Sequence like a calcium-activated K+ channel;SLACK;Slo2.2
- bA100C15.2;EIEE14;ENFL5;KCa4.1;KCNT1;KCNT1_HUMAN;Potassium channel subfamily T member 1;Potassium channel, sodium activated subfamily T, member 1;Potassium channel, subfamily T, member 1;Sequence like a calcium-activated K+ channel;SLACK;Slo2.2
- Antigen Name:
- Potassium channel subfamily T member 1 protein (1026-1230AA)
- Potassium channel subfamily T member 1 protein (1026-1230AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5JUK3
- Q5JUK3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together