Anti-CSPG4 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:20 to 1:100
Type:
Antigen: CSPG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77820-690
- 77820-689
- Antigen Symbol:
- CSPG4
- CSPG4
- Antigen Name:
- Synthetic peptide of CSPG4
- Synthetic peptide of CSPG4
- Antigen Synonyms:
- 4732461B14Rik|AN2|AN2 proteoglycan|Chondroitin sulfate proteoglycan 4 (melanoma-associated)|Chondroitin sulfate proteoglycan 4|Chondroitin sulfate proteoglycan NG2|Cspg4|Cspg4 chondroitin sulfate proteoglycan 4|CSPG4_HUMAN|HMW-MAA|HSN tumor-specific antigen|Kiaa4232|MCSP|MCSPG|MEL-CSPG|Melanoma chondroitin sulfate proteoglycan|Melanoma-associated chondroitin sulfate proteoglycan|MELCSPG|MSK16|NG2
- 4732461B14Rik|AN2|AN2 proteoglycan|Chondroitin sulfate proteoglycan 4 (melanoma-associated)|Chondroitin sulfate proteoglycan 4|Chondroitin sulfate proteoglycan NG2|Cspg4|Cspg4 chondroitin sulfate proteoglycan 4|CSPG4_HUMAN|HMW-MAA|HSN tumor-specific antigen|Kiaa4232|MCSP|MCSPG|MEL-CSPG|Melanoma chondroitin sulfate proteoglycan|Melanoma-associated chondroitin sulfate proteoglycan|MELCSPG|MSK16|NG2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6UVK1
- Q6UVK1
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together