Anti-NDUFAB1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: NDUFAB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse
Specifications
- Catalog No:
- 77820-938
- 77820-939
- Antigen Symbol:
- NDUFAB1
- NDUFAB1
- Antigen Name:
- Synthetic peptide of NDUFAB1
- Synthetic peptide of NDUFAB1
- Antigen Synonyms:
- ACP|ACPM_HUMAN|Acyl carrier protein|Acyl carrier protein mitochondrial|CI SDAP|CI-SDAP|FASN 2A|FASN2A|MGC65095|Mitochondrial acyl carrier protein|mitochondrial|NADH dehydrogenase (ubiquinone) 1 alpha/beta subcomplex 1 8kDa|NADH dehydrogenase (ubiquinone) 1 alpha/beta subcomplex 1|NADH ubiquinone oxidoreductase 9.6 kDa subunit|NADH ubiquinone oxidoreductase SDAP subunit|NADH-ubiquinone oxidoreductase 9.6 kDa subunit|NDUFAB 1|ndufab1|SDAP
- ACP|ACPM_HUMAN|Acyl carrier protein|Acyl carrier protein mitochondrial|CI SDAP|CI-SDAP|FASN 2A|FASN2A|MGC65095|Mitochondrial acyl carrier protein|mitochondrial|NADH dehydrogenase (ubiquinone) 1 alpha/beta subcomplex 1 8kDa|NADH dehydrogenase (ubiquinone) 1 alpha/beta subcomplex 1|NADH ubiquinone oxidoreductase 9.6 kDa subunit|NADH ubiquinone oxidoreductase SDAP subunit|NADH-ubiquinone oxidoreductase 9.6 kDa subunit|NDUFAB 1|ndufab1|SDAP
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14561
- O14561
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together