Anti-POMC Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000
Type:
Antigen: POMC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77812-819
- 77812-820
- Antigen Symbol:
- POMC
- POMC
- Antigen Name:
- Synthetic peptide of POMC
- Synthetic peptide of POMC
- Antigen Synonyms:
- ACTH|Adrenocorticotropic hormone|Adrenocorticotropin|alpha melanocyte stimulating hormone|Alpha MSH|Alpha-MSH|beta endorphin|Beta LPH|beta melanocyte stimulating hormone|Beta MSH|Beta-LPH|Beta-MSH|CLIP|COLI_HUMAN|Corticotropin like intermediary peptide|Corticotropin lipotropin|Corticotropin-lipotropin|Gamma MSH|Gamma-LPH|Gamma-MSH|Lipotropin beta|Lipotropin gamma|LPH|Melanotropin alpha|Melanotropin|Melanotropin beta|Melanotropin gamma|met enkephalin|Met-enkephalin|MSH|NPP|POC|pro ACTH endorphin|Proopiomelanocortin|Proopiomelanocortin preproprotein
- ACTH|Adrenocorticotropic hormone|Adrenocorticotropin|alpha melanocyte stimulating hormone|Alpha MSH|Alpha-MSH|beta endorphin|Beta LPH|beta melanocyte stimulating hormone|Beta MSH|Beta-LPH|Beta-MSH|CLIP|COLI_HUMAN|Corticotropin like intermediary peptide|Corticotropin lipotropin|Corticotropin-lipotropin|Gamma MSH|Gamma-LPH|Gamma-MSH|Lipotropin beta|Lipotropin gamma|LPH|Melanotropin alpha|Melanotropin|Melanotropin beta|Melanotropin gamma|met enkephalin|Met-enkephalin|MSH|NPP|POC|pro ACTH endorphin|Proopiomelanocortin|Proopiomelanocortin preproprotein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P01189
- P01189
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together