Anti-PTGES2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: PTGES2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77818-444
- 77818-445
- Antigen Symbol:
- PTGES2
- PTGES2
- Antigen Name:
- Fusion protein of PTGES2
- Fusion protein of PTGES2
- Antigen Synonyms:
- C9orf15|FLJ14038|Gamma interferon activated transcriptional element binding factor 1|GATE binding factor 1|GBF 1|GBF1|Membrane associated prostaglandin E synthase 2|MGC11289|Microsomal prostaglandin E synthase 2|mPGES 2|mPGES-2|PGES2|PGES2_HUMAN|Prostaglandin E synthase 2|Prostaglandin E synthase 2 truncated form|PTGES 2|PTGES2
- C9orf15|FLJ14038|Gamma interferon activated transcriptional element binding factor 1|GATE binding factor 1|GBF 1|GBF1|Membrane associated prostaglandin E synthase 2|MGC11289|Microsomal prostaglandin E synthase 2|mPGES 2|mPGES-2|PGES2|PGES2_HUMAN|Prostaglandin E synthase 2|Prostaglandin E synthase 2 truncated form|PTGES 2|PTGES2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H7Z7
- Q9H7Z7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together