Anti-TMPRSS3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: TMPRSS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Specifications
- Catalog No:
- 77808-906
- 77808-907
- Antigen Symbol:
- TMPRSS3
- TMPRSS3
- Antigen Name:
- Synthetic peptide of TMPRSS3
- Synthetic peptide of TMPRSS3
- Antigen Synonyms:
- Deafness autosomal recessive 10| DFNB10| DFNB8| ECHOS1| Gene similar to transmembrane serine protease| MGC130589| Serine protease TADG-12| si:dz69g10.3| TADG12| TMPRSS 3| TMPRSS3| TMPS3_HUMAN| Transmembrane protease serine 3| Tumor associated differentially expressed gene 12 protein| Tumor-associated differentially-expressed gene 12 protein| UNQ323/PRO382
- Deafness autosomal recessive 10| DFNB10| DFNB8| ECHOS1| Gene similar to transmembrane serine protease| MGC130589| Serine protease TADG-12| si:dz69g10.3| TADG12| TMPRSS 3| TMPRSS3| TMPS3_HUMAN| Transmembrane protease serine 3| Tumor associated differentially expressed gene 12 protein| Tumor-associated differentially-expressed gene 12 protein| UNQ323/PRO382
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P57727
- P57727
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together