Anti-NFIL3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NFIL3
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-620
- 77824-619
- Antigen Symbol:
- NFIL3
- NFIL3
- Antigen Name:
- Nuclear factor interleukin-3-regulated protein (133-462AA)
- Nuclear factor interleukin-3-regulated protein (133-462AA)
- Antigen Synonyms:
- E4 promoter binding protein 4| E4 promoter-binding protein 4| E4BP 4| E4BP4| E4BP4 protein| IL3BP 1| IL3BP1| Interleukin 3 binding protein 1 | Interleukin 3 promoter transcriptional activator| Interleukin-3 promoter transcriptional activator| Interleukin-3-binding protein 1| NFIL 3| NFIL 3A| NFIL3| NFIL3_HUMAN| NFIL3A| Nuclear factor interleukin 3 regulated| Nuclear factor interleukin-3-regulated protein| Transcriptional activator NF IL3A| Transcriptional activator NF-IL3A
- E4 promoter binding protein 4| E4 promoter-binding protein 4| E4BP 4| E4BP4| E4BP4 protein| IL3BP 1| IL3BP1| Interleukin 3 binding protein 1 | Interleukin 3 promoter transcriptional activator| Interleukin-3 promoter transcriptional activator| Interleukin-3-binding protein 1| NFIL 3| NFIL 3A| NFIL3| NFIL3_HUMAN| NFIL3A| Nuclear factor interleukin 3 regulated| Nuclear factor interleukin-3-regulated protein| Transcriptional activator NF IL3A| Transcriptional activator NF-IL3A
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16649
- Q16649
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together