Anti-ZNF644 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: ZNF644
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77805-745
- 77805-744
- Antigen Symbol:
- ZNF644
- ZNF644
- Antigen Name:
- Zinc finger protein 644 protein (752-913AA)
- Zinc finger protein 644 protein (752-913AA)
- Antigen Synonyms:
- BM005|KIAA1221|RGD1306805|Zep-2|Zep2|Zinc finger motif enhancer binding protein 2|Zinc finger motif enhancer-binding protein 2|Zinc finger protein 644|Zinc finger protein 644, isoform 2|ZN644_HUMAN|ZNF644
- BM005|KIAA1221|RGD1306805|Zep-2|Zep2|Zinc finger motif enhancer binding protein 2|Zinc finger motif enhancer-binding protein 2|Zinc finger protein 644|Zinc finger protein 644, isoform 2|ZN644_HUMAN|ZNF644
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H582
- Q9H582
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together