Anti-ZFAND3 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC:1:20 to 1:200
Type:
Antigen: ZFAND3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77837-247
- 77837-246
- Antigen Symbol:
- AN1-type zinc finger protein 3;Testis expressed sequence 27;Testis-expressed sequence 27;TEX27;ZFAN3_HUMAN;Zfand3;Zinc finger AN1type domain 3
- AN1-type zinc finger protein 3;Testis expressed sequence 27;Testis-expressed sequence 27;TEX27;ZFAN3_HUMAN;Zfand3;Zinc finger AN1type domain 3
- Antigen Name:
- AN1-type zinc finger protein 3 protein (1-227AA)
- AN1-type zinc finger protein 3 protein (1-227AA)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H8U3
- Q9H8U3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together