Anti-UBE2L6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2L6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-869
- 77803-869
- Antigen Symbol:
- UBE2L6
- UBE2L6
- Antigen Name:
- Ubiquitin/ISG15-conjugating enzyme E2 L6 protein (13-153AA)
- Ubiquitin/ISG15-conjugating enzyme E2 L6 protein (13-153AA)
- Antigen Synonyms:
- MGC40331|Retinoic acid induced gene B protein|Retinoic acid-induced gene B protein|RIG B|RIG-B|UB2L6_HUMAN|UBCH 8|UbcH8|UBE2L6|Ubiquitin carrier protein|Ubiquitin carrier protein L6|Ubiquitin conjugating enzyme E2L 6|Ubiquitin protein ligase|Ubiquitin protein ligase L6|Ubiquitin-protein ligase L6|Ubiquitin/ISG15 conjugating enzyme E2 L6|Ubiquitin/ISG15-conjugating enzyme E2 L6
- MGC40331|Retinoic acid induced gene B protein|Retinoic acid-induced gene B protein|RIG B|RIG-B|UB2L6_HUMAN|UBCH 8|UbcH8|UBE2L6|Ubiquitin carrier protein|Ubiquitin carrier protein L6|Ubiquitin conjugating enzyme E2L 6|Ubiquitin protein ligase|Ubiquitin protein ligase L6|Ubiquitin-protein ligase L6|Ubiquitin/ISG15 conjugating enzyme E2 L6|Ubiquitin/ISG15-conjugating enzyme E2 L6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O14933
- O14933
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together