Anti-TCF15 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TCF15
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77823-811
- 77823-810
- Antigen Symbol:
- TCF15
- TCF15
- Antigen Name:
- Transcription factor 15 protein (24-199AA)
- Transcription factor 15 protein (24-199AA)
- Antigen Synonyms:
- bHLH EC2 protein|bHLHa40|BHLHEC2|Class A basic helix-loop-helix protein 40|EC2|Paraxis|Protein bHLH-EC2|TCF-15|Tcf15|TCF15_HUMAN|Transcription factor 15
- bHLH EC2 protein|bHLHa40|BHLHEC2|Class A basic helix-loop-helix protein 40|EC2|Paraxis|Protein bHLH-EC2|TCF-15|Tcf15|TCF15_HUMAN|Transcription factor 15
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12870
- Q12870
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together